siRNA Details
| Gene Symbol: Gli4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029926.1 | NP_001025097.1 | 4 | 1274 | ||
| XM_006241856.2 | XP_006241918.1 | 4 | 873 | |||
| XM_006241858.1 | XP_006241920.1 | 4 | 1158 | |||
| GenBank mRNA | BC097383.1 | - | 4 | 1274 | 7446306 | |
| Gene Symbol: LOC100911986 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752188.3 | XP_003752236.2 | 1 | 991 | ||

Gli4
Genomic Map