siRNA Details
| Gene Symbol: Ncf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127304.1 | NP_001120776.1 | 3 | 366 | ||
| XM_006241939.2 | XP_006242001.1 | 4 | 1030 | |||
| GenBank mRNA | BC167076.1 | - | 3 | 338 | 7129758 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncf4| Gene Symbol: Ncf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127304.1 | NP_001120776.1 | 3 | 366 | ||
| XM_006241939.2 | XP_006242001.1 | 4 | 1030 | |||
| GenBank mRNA | BC167076.1 | - | 3 | 338 | 7129758 | |