siRNA Details
| Gene Symbol: Ccdc134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024355.1 | NP_001019526.1 | 4 | 595 | ||
| XM_006242113.2 | XP_006242175.1 | 5 | 508 | |||
| GenBank mRNA | BC088206.1 | - | 4 | 595 | 7381354 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc134| Gene Symbol: Ccdc134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024355.1 | NP_001019526.1 | 4 | 595 | ||
| XM_006242113.2 | XP_006242175.1 | 5 | 508 | |||
| GenBank mRNA | BC088206.1 | - | 4 | 595 | 7381354 | |