siRNA Details
| Gene Symbol: RGD1566029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241417.2 | XP_006241479.1 | 3 | 613 | ||
| XM_008765816.1 | XP_008764038.1 | 3 | 614 | |||
| XM_008765817.1 | XP_008764039.1 | 3 | 614 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1566029| Gene Symbol: RGD1566029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241417.2 | XP_006241479.1 | 3 | 613 | ||
| XM_008765816.1 | XP_008764038.1 | 3 | 614 | |||
| XM_008765817.1 | XP_008764039.1 | 3 | 614 | |||