siRNA Details
| Gene Symbol: Odf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191992.1 | NP_001178921.1 | 4 | 478 | ||
| XM_006242237.1 | XP_006242299.1 | 4 | 587 | |||
| XM_006242238.2 | XP_006242300.1 | 5 | 1121 | |||
| XM_008765776.1 | XP_008763998.1 | 4 | 861 | |||

Odf3b
Genomic Map