siRNA Details
| Gene Symbol: Tmem117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271239.1 | NP_001258168.1 | 8 | 1127 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem117| Gene Symbol: Tmem117 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271239.1 | NP_001258168.1 | 8 | 1127 | ||