siRNA Details
| Gene Symbol: Or10ad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765883.1 | XP_008764105.1 | 1 | 379 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Or10ad1| Gene Symbol: Or10ad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765883.1 | XP_008764105.1 | 1 | 379 | ||