siRNA Details
| Gene Symbol: Olr1877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242360.2 | XP_006242422.1 | 3 | 647 | ||
| XM_008765852.1 | XP_008764074.1 | 2 | 426 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1877| Gene Symbol: Olr1877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242360.2 | XP_006242422.1 | 3 | 647 | ||
| XM_008765852.1 | XP_008764074.1 | 2 | 426 | |||