siRNA Details
| Gene Symbol: Olr1877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242360.2 | XP_006242422.1 | 3 | 999 | ||
| XM_008765852.1 | XP_008764074.1 | 2 | 778 | |||
| Gene Symbol: Olr1102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765851.1 | XP_008764073.1 | 2 | 814 | ||

Olr1877
Genomic Map