siRNA Details
| Gene Symbol: Tuba1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044270.2 | NP_001037735.1 | 4 | 1598 | ||
| GenBank mRNA | BC060572.1 | - | 4 | 1602 | 6920989 | |
| BC076379.1 | - | 4 | 1584 | 7106502 | ||
| FQ212426.1 | - | 4 | 1593 | |||
| FQ212740.1 | - | 4 | 1593 | |||
| FQ213098.1 | - | 4 | 1598 | |||
| FQ213226.1 | - | 4 | 1596 | |||
| FQ213439.1 | - | 4 | 1595 | |||
| FQ213947.1 | - | 4 | 1594 | |||
| FQ214075.1 | - | 4 | 1596 | |||
| FQ214100.1 | - | 4 | 1593 | |||
| FQ214301.1 | - | 4 | 1597 | |||
| FQ223081.1 | - | 4 | 1598 | |||
| FQ225046.1 | - | 4 | 1602 | |||
| FQ225339.1 | - | 4 | 1592 | |||
| FQ228903.1 | - | 1381 | ||||
| FQ231550.1 | - | 4 | 1594 | |||
| FQ232160.1 | - | 3 | 1503 | |||
| FQ232527.1 | - | 4 | 1598 | |||
| FQ233000.1 | - | 4 | 1595 | |||
| FQ233566.1 | - | 4 | 1596 | |||
| FQ234135.1 | - | 4 | 1593 | |||
| V01226.1 | - | 1 | 581 | |||

Tuba1b
Genomic Map