siRNA Details
| Gene Symbol: Arhgap42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242515.2 | XP_006242577.1 | 9 | 1640 | ||
| XM_008765909.1 | XP_008764131.1 | 9 | 2093 | |||
| XM_576354.6 | XP_576354.3 | 9 | 1183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap42| Gene Symbol: Arhgap42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242515.2 | XP_006242577.1 | 9 | 1640 | ||
| XM_008765909.1 | XP_008764131.1 | 9 | 2093 | |||
| XM_576354.6 | XP_576354.3 | 9 | 1183 | |||