siRNA Details
Gene Symbol: RGD1561795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109289.1 | NP_001102759.1 | 4 | 614 | ||
XM_008765967.1 | XP_008764189.1 | 6 | 953 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1561795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109289.1 | NP_001102759.1 | 4 | 614 | ||
XM_008765967.1 | XP_008764189.1 | 6 | 953 |