siRNA Details
| Gene Symbol: LOC500948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047964.1 | NP_001041429.1 | 541 | |||
| GenBank mRNA | AY387059.1 | - | 541 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC500948| Gene Symbol: LOC500948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047964.1 | NP_001041429.1 | 541 | |||
| GenBank mRNA | AY387059.1 | - | 541 | |||