siRNA Details
| Gene Symbol: Zfp317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134634.2 | NP_001128106.2 | 9 | 1021 | ||
| XM_006242578.2 | XP_006242640.1 | 8 | 1013 | |||
| XM_008765969.1 | XP_008764191.1 | 7 | 827 | |||
| XM_008765970.1 | XP_008764192.1 | 8 | 888 | |||
| XM_008765971.1 | XP_008764193.1 | 7 | 831 | |||
| XM_008765972.1 | XP_008764194.1 | 8 | 892 | |||
| XM_008765973.1 | XP_008764195.1 | 6 | 769 | |||
| XM_008765974.1 | XP_008764196.1 | 6 | 773 | |||
| XM_008765975.1 | XP_008764197.1 | 7 | 803 | |||
| XM_008765976.1 | XP_008764198.1 | 6 | 665 | |||
| XM_008765977.1 | XP_008764199.1 | 6 | 641 | |||
| GenBank mRNA | BU671127.1 | - | 5 | 391 | ||
| FQ211838.1 | - | 7 | 808 | |||

Zfp317
Genomic Map