siRNA Details
| Gene Symbol: Ubl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048243.2 | NP_001041708.2 | 3 | 238 | ||
| XM_006242661.2 | XP_006242723.1 | 3 | 419 | |||
| GenBank mRNA | BC091358.1 | - | 2 | 432 | ||
| BC157818.1 | - | 2 | 141 | 7936655 | ||
| CB713235.1 | - | 3 | 238 | |||

Ubl5
Genomic Map