siRNA Details
| Gene Symbol: GRAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291824.1 | NP_001278753.1 | 5 | 589 | ||
| NM_001291825.1 | NP_001278754.1 | 4 | 602 | |||
| NM_001291826.1 | NP_001278755.1 | 4 | 497 | |||
| NM_004810.3 | NP_004801.1 | 5 | 575 | |||
| XM_006724376.3 | XP_006724439.1 | 4 | 492 | |||
| GenBank mRNA | AF042380.1 | - | 5 | 504 | ||
| AF090456.1 | - | 5 | 466 | |||
| AF102694.1 | - | 5 | 504 | |||
| AF121002.1 | - | 4 | 609 | |||
| AF129476.1 | - | 5 | 494 | |||
| AF236119.1 | - | 5 | 535 | |||
| AF236120.1 | - | 4 | 312 | |||
| AJ011736.1 | - | 5 | 520 | |||
| AK303280.1 | - | 4 | 482 | |||
| AK303332.1 | - | 3 | 389 | |||
| AK303512.1 | - | 2 | 244 | |||
| AK303657.1 | - | 4 | 454 | |||
| AK312830.1 | - | 5 | 574 | |||
| AY069959.1 | - | 5 | 575 | |||
| BC025692.1 | - | 5 | 509 | 5208327 | ||
| BC026002.1 | - | 4 | 571 | 4672797 | ||
| CR456498.1 | - | 5 | 373 | |||
| CR536524.1 | - | 4 | 312 | |||
| CR541849.1 | - | 4 | 312 | |||
| Y18051.1 | - | 5 | 348 | |||

GRAP2
Genomic Map