siRNA Details
| Gene Symbol: Ddx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109292.1 | NP_001102762.1 | 10 | 1443 | ||
| XM_006242950.2 | XP_006243012.1 | 10 | 1443 | |||
| XM_006242951.2 | XP_006243013.1 | 10 | 1457 | |||
| XM_006242952.2 | XP_006243014.1 | 10 | 1452 | |||
| XM_006242954.2 | XP_006243016.1 | 10 | 1726 | |||
| GenBank mRNA | FQ234306.1 | - | 2 | 238 | ||
| FQ235180.1 | - | 2 | 228 | |||

Ddx6
Genomic Map