siRNA Details
| Gene Symbol: Zfp259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137646.1 | NP_001131118.1 | 5 | 542 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp259| Gene Symbol: Zfp259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137646.1 | NP_001131118.1 | 5 | 542 | ||