siRNA Details
| Gene Symbol: Layn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191997.1 | NP_001178926.1 | 7 | 689 | ||
| XM_006243049.2 | XP_006243111.1 | 6 | 1654 | |||
| XM_006243052.2 | XP_006243114.1 | 5 | 538 | |||
| XM_008766328.1 | XP_008764550.1 | 5 | 1618 | |||

Layn
Genomic Map