siRNA Details
| Gene Symbol: Spesp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077682.1 | NP_001071150.1 | 2 | 1171 | ||
| XM_006243210.2 | XP_006243272.1 | 2 | 1235 | |||
| GenBank mRNA | BC127527.1 | - | 2 | 1171 | 7114748 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spesp1| Gene Symbol: Spesp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077682.1 | NP_001071150.1 | 2 | 1171 | ||
| XM_006243210.2 | XP_006243272.1 | 2 | 1235 | |||
| GenBank mRNA | BC127527.1 | - | 2 | 1171 | 7114748 | |