siRNA Details
Gene Symbol: Spesp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077682.1 | NP_001071150.1 | 2 | 1207 | ||
XM_006243210.2 | XP_006243272.1 | 2 | 1271 | |||
GenBank mRNA | BC127527.1 | - | 2 | 1207 | 7114748 |
