siRNA Details
| Gene Symbol: Gfral | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191998.1 | NP_001178927.1 | 6 | 807 | ||
| XM_006243416.2 | XP_006243478.1 | 7 | 2847 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfral| Gene Symbol: Gfral | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191998.1 | NP_001178927.1 | 6 | 807 | ||
| XM_006243416.2 | XP_006243478.1 | 7 | 2847 | |||