siRNA Details
| Gene Symbol: Sh3bgrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137647.1 | NP_001131119.1 | 2 | 295 | ||
| Gene Symbol: LOC102554622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243468.2 | XP_006243530.1 | 2 | 290 | ||

Sh3bgrl2
Genomic Map