siRNA Details
| Gene Symbol: LOC501038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047965.1 | NP_001041430.1 | 2 | 131 | ||
| XM_006243552.2 | XP_006243614.1 | 2 | 1658 | |||
| GenBank mRNA | AY325184.1 | - | 2 | 131 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC501038| Gene Symbol: LOC501038 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047965.1 | NP_001041430.1 | 2 | 131 | ||
| XM_006243552.2 | XP_006243614.1 | 2 | 1658 | |||
| GenBank mRNA | AY325184.1 | - | 2 | 131 | ||