siRNA Details
| Gene Symbol: Slc25a36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766577.1 | XP_008764799.1 | 6 | 1806 | ||
| XM_576451.6 | XP_576451.4 | 7 | 1883 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a36| Gene Symbol: Slc25a36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766577.1 | XP_008764799.1 | 6 | 1806 | ||
| XM_576451.6 | XP_576451.4 | 7 | 1883 | |||