siRNA Details
| Gene Symbol: Rps23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | FQ221811.1 | - | 2 | 182 | ||
| FQ222494.1 | - | 2 | 184 | |||
| FQ222496.1 | - | 2 | 181 | |||
| FQ222641.1 | - | 2 | 182 | |||
| FQ223677.1 | - | 2 | 182 | |||
| FQ223812.1 | - | 2 | 182 | |||
| FQ228659.1 | - | 2 | 182 | |||
| FQ228931.1 | - | 2 | 182 | |||

Rps23
Genomic Map