siRNA Details
| Gene Symbol: RGD1559960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244256.2 | XP_006244318.2 | 5 | 669 | ||
| XM_008766792.1 | XP_008765014.1 | 6 | 816 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1559960| Gene Symbol: RGD1559960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244256.2 | XP_006244318.2 | 5 | 669 | ||
| XM_008766792.1 | XP_008765014.1 | 6 | 816 | |||