siRNA Details
| Gene Symbol: Treml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192001.1 | NP_001178930.1 | 5 | 695 | ||
| GenBank mRNA | FQ227710.1 | - | 5 | 705 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Treml1| Gene Symbol: Treml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192001.1 | NP_001178930.1 | 5 | 695 | ||
| GenBank mRNA | FQ227710.1 | - | 5 | 705 | ||