siRNA Details
| Gene Symbol: Mdfi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109301.1 | NP_001102771.1 | 5 | 1002 | ||
| XM_006244475.2 | XP_006244537.1 | 5 | 763 | |||
| XM_006244476.2 | XP_006244538.1 | 5 | 956 | |||
| XM_008766877.1 | XP_008765099.1 | 5 | 929 | |||
| GenBank mRNA | BC158727.1 | - | 4 | 924 | 9091612 | |

Mdfi
Genomic Map