siRNA Details
| Gene Symbol: Lrrc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024360.2 | NP_001019531.1 | 5 | 830 | ||
| XM_008766878.1 | XP_008765100.1 | 4 | 1806 | |||
| GenBank mRNA | AB210855.1 | - | 5 | 730 | ||
| BC128712.1 | - | 5 | 830 | 7133504 | ||

Lrrc73
Genomic Map