siRNA Details
Gene Symbol: Slc35b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037215.3 | NP_001032292.1 | 4 | 853 | ||
GenBank mRNA | AB197928.1 | - | 4 | 803 | ||
BC100647.1 | - | 4 | 853 | 7932661 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc35b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037215.3 | NP_001032292.1 | 4 | 853 | ||
GenBank mRNA | AB197928.1 | - | 4 | 803 | ||
BC100647.1 | - | 4 | 853 | 7932661 |