siRNA Details
| Gene Symbol: Gpr115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244640.2 | XP_006244702.1 | 6 | 962 | ||
| GenBank mRNA | FQ222190.1 | - | 6 | 962 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr115| Gene Symbol: Gpr115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244640.2 | XP_006244702.1 | 6 | 962 | ||
| GenBank mRNA | FQ222190.1 | - | 6 | 962 | ||