siRNA Details
Gene Symbol: Gpr115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244640.2 | XP_006244702.1 | 6 | 1813 | ||
GenBank mRNA | FQ222190.1 | - | 6 | 1813 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpr115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244640.2 | XP_006244702.1 | 6 | 1813 | ||
GenBank mRNA | FQ222190.1 | - | 6 | 1813 |