siRNA Details
| Gene Symbol: Amer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244725.2 | XP_006244787.1 | 3 | 2382 | ||
| XM_576539.5 | XP_576539.3 | 2 | 2281 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amer3| Gene Symbol: Amer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244725.2 | XP_006244787.1 | 3 | 2382 | ||
| XM_576539.5 | XP_576539.3 | 2 | 2281 | |||