siRNA Details
| Gene Symbol: Fam178b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122658.1 | NP_001116130.1 | 3 | 608 | ||
| XM_006244842.2 | XP_006244904.1 | 3 | 629 | |||
| XM_006244843.2 | XP_006244905.1 | 4 | 743 | |||
| XM_006244844.2 | XP_006244906.1 | 4 | 700 | |||
| XM_008767041.1 | XP_008765263.1 | 2 | 375 | |||
| GenBank mRNA | BC160833.1 | - | 3 | 608 | 7456084 | |

Fam178b
Genomic Map