siRNA Details
| Gene Symbol: FAM189A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127608.1 | NP_001121080.1 | 10 | 1080 | ||
| NM_004816.3 | NP_004807.3 | 10 | 1396 | |||
| XM_005252307.4 | XP_005252364.1 | 10 | 1957 | |||
| XM_017015324.1 | XP_016870813.1 | 9 | 1141 | |||
| GenBank mRNA | AK056152.1 | - | 8 | 1024 | ||
| AK126127.1 | - | 10 | 1396 | |||
| BC113683.1 | - | 10 | 1039 | 8322735 | ||
| BC113685.1 | - | 10 | 1039 | 8322737 | ||
| BX641153.1 | - | 6 | 8461 | |||

FAM189A2
Genomic Map