siRNA Details
| Gene Symbol: Satb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109306.1 | NP_001102776.1 | 4 | 718 | ||
| XM_006244931.2 | XP_006244993.1 | 4 | 1390 | |||
| XM_008767128.1 | XP_008765350.1 | 3 | 1162 | |||
| XM_008767129.1 | XP_008765351.1 | 4 | 664 | |||

Satb2
Genomic Map