siRNA Details
| Gene Symbol: RGD1562029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767174.1 | XP_008765396.1 | 7 | 1604 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562029| Gene Symbol: RGD1562029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767174.1 | XP_008765396.1 | 7 | 1604 | ||