siRNA Details
| Gene Symbol: Rpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033903.1 | NP_001029075.1 | 1, 2 | 136 | ||
| XM_006245104.2 | XP_006245166.1 | 1, 2 | 232 | |||
| XM_006245105.2 | XP_006245167.1 | 1, 2 | 235 | |||
| XM_006245106.2 | XP_006245168.1 | 1, 2 | 231 | |||
| XM_006245107.2 | XP_006245169.1 | 1, 2 | 232 | |||
| XM_006245109.2 | XP_006245171.1 | 1, 2 | 228 | |||
| XM_008767287.1 | XP_008765509.1 | 1, 2 | 236 | |||
| XM_008767288.1 | XP_008765510.1 | 1, 2 | 240 | |||
| GenBank mRNA | BC101869.1 | - | 1, 2 | 136 | 5621971 | |

Rpe
Genomic Map