siRNA Details
Gene Symbol: Mreg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192002.1 | NP_001178931.1 | 4 | 401 | ||
XM_006245265.2 | XP_006245327.1 | 5 | 549 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mreg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192002.1 | NP_001178931.1 | 4 | 401 | ||
XM_006245265.2 | XP_006245327.1 | 5 | 549 |