siRNA Details
| Gene Symbol: LOC501180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024364.1 | NP_001019535.1 | 1 | 110 | ||
| GenBank mRNA | BC085806.1 | - | 1 | 110 | 7112277 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC501180| Gene Symbol: LOC501180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024364.1 | NP_001019535.1 | 1 | 110 | ||
| GenBank mRNA | BC085806.1 | - | 1 | 110 | 7112277 | |