siRNA Details
| Gene Symbol: St8sia4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053914.1 | NP_446366.1 | 1 | 339 | ||
| GenBank mRNA | AM419015.1 | - | 1 | 339 | ||
| U90215.1 | - | 1 | 65 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St8sia4| Gene Symbol: St8sia4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053914.1 | NP_446366.1 | 1 | 339 | ||
| GenBank mRNA | AM419015.1 | - | 1 | 339 | ||
| U90215.1 | - | 1 | 65 | |||