siRNA Details
| Gene Symbol: Ppid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004279.1 | NP_001004279.1 | 8 | 988 | ||
| GenBank mRNA | BC076386.1 | - | 8 | 988 | 7108860 | |
| FQ214826.1 | - | 8 | 991 | |||
| Gene Symbol: Ppidl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750757.3 | XP_003750805.1 | 1 | 936 | ||

Ppid
Genomic Map