siRNA Details
Gene Symbol: LOC501391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227951.2 | XP_006228013.1 | 4 | 483 | ||
XM_008758800.1 | XP_008757022.1 | 4 | 868 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC501391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227951.2 | XP_006228013.1 | 4 | 483 | ||
XM_008758800.1 | XP_008757022.1 | 4 | 868 |