siRNA Details
| Gene Symbol: Sh2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109313.2 | NP_001102783.2 | 3 | 472 | ||
| XM_008773527.1 | XP_008771749.1 | 3 | 611 | |||
| XM_008773528.1 | XP_008771750.1 | 3 | 602 | |||
| GenBank mRNA | BC167036.1 | - | 3 | 472 | 7389183 | |

Sh2d1a
Genomic Map