siRNA Details
| Gene Symbol: RGD1560927 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109315.1 | NP_001102785.1 | 2 | 386 | ||
| XM_006257482.1 | XP_006257544.1 | 2 | 291 | |||
| GenBank mRNA | FQ221578.1 | - | 2 | 284 | ||
| Gene Symbol: LOC103694509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773491.1 | XP_008771713.1 | 2 | 405 | ||

RGD1560927
Genomic Map