siRNA Details
| Gene Symbol: RGD1559731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773060.1 | XP_008771282.1 | 3 | 527 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1559731| Gene Symbol: RGD1559731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773060.1 | XP_008771282.1 | 3 | 527 | ||