siRNA Details
| Gene Symbol: Zcchc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253531.1 | XP_006253593.1 | 20 | 3891 | ||
| XM_006253532.2 | XP_006253594.1 | 20 | 3742 | |||
| XM_006253533.1 | XP_006253595.1 | 20 | 3649 | |||
| XM_008771464.1 | XP_008769686.1 | 20 | 3742 | |||
| XM_008771465.1 | XP_008769687.1 | 20 | 3741 | |||
| XM_008771466.1 | XP_008769688.1 | 20 | 3741 | |||

Zcchc6
Genomic Map