siRNA Details
| Gene Symbol: Ammecr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002730211.4 | XP_002730257.1 | 6 | 995 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ammecr1| Gene Symbol: Ammecr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002730211.4 | XP_002730257.1 | 6 | 995 | ||