siRNA Details
| Gene Symbol: Fancb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001192003.1 | NP_001178932.1 | 5 | 1474 | ||
| XM_006256884.2 | XP_006256946.1 | 6 | 1897 | |||
| XM_006256885.2 | XP_006256947.1 | 7 | 1937 | |||
| XM_006256886.2 | XP_006256948.1 | 6 | 1899 | |||
| XM_008773184.1 | XP_008771406.1 | 6 | 1898 | |||
| XM_008773185.1 | XP_008771407.1 | 8 | 2041 | |||

Fancb
Genomic Map